Learn More
GPE Scientific™ J Young Schlenk Flask, Cylindrical with Fine Thread Tap and Cone
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Pakningsstørrelse:
Each
Beskrivelse
- Solidly constructed for use with vacuum manifolds
- Designed for use with Schlenk lines
- Includes fine thread tap and cone
Tekniske data
Tekniske data
| Capacity (Metric) | 1000 mL |
| For Use With (Equipment) | Vacuum manifolds and Schlenk lines |
| Includes | Fine thread tap and cone |
| Product Type | Schlenk Flask |
| Socket Joint | Size: 29 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion