Learn More
GPE Scientific™ J Young Spring Clips for Spherical Joints
GPE Scientific™ J Young Spring Clips for Spherical Joints are spring-based clips designed to securely fasten J Young spherical joints.
19.21€ - 79.60€
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
15894774
|
GPE Scientific™
JC13 |
12/2 and 12/5 spherical joints |
19.30€
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||||
15804784
|
GPE Scientific™
JC19 |
18/7 and 18/9 spherical joints |
27.35€
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||||
15834784
|
GPE Scientific™
JC41 |
40/25 spherical joints |
26.85€
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||||
15844784
|
GPE Scientific™
JC51 |
51/35 spherical joints |
79.60€
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
15814784
|
GPE Scientific™
JC29 |
28/12 and 28/15 spherical joints |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15824784
|
GPE Scientific™
JC35 |
35/20 spherical joints |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Description
- Spring clip design
- Securely fasten J Young spherical joints
Specifications
Spring Clips for J Young Spherical Joints |