Learn More
Emma Safety Footwear Jack (Ruffneck) Safety Shoes
Sneaker with composite toe cap includes a unique rough neck heel design which is highly supportive and ideal for climbing pallets, ladders and getting in and out of truck cabs even when wet. The virtually impenetrable insole protects from sharp objects like nails and shards of metal.
Brand:  Emma Safety Footwear 912549-43
Additional Details : Weight : 1.56800kg
 View more versions of this product
Product Code. 15949061
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • S1P-Safety shoe with sole plate for dry work
  • Denim/Nubuck construction
  • GripForce™ Easy Twist Outsole
  • 3M™ Reflective Striping
  • ESD certified
  • Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifications
Specifications
EN ISO 20345:2011
Blue
Hydro-Tec™ Sanitized Silver
43
Laces
Jack (Ruffneck)
Denim, Nubuck
Composite