missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimberly-Clark Professional™ KleenGuard™ Jackson Safety™ G80 Chemical Resistant Nitrile Gloves
99.12 EUR valid until 2024-12-31
Use promo code "21907" to get your promotional price.
84.25 EUR valid until 2024-04-30
Use promo code "21673" to get your promotional price.
Use promo code "21907" to get your promotional price.
84.25 EUR valid until 2024-04-30
Use promo code "21673" to get your promotional price.
missing translation for 'orderingAttributeHoverText'
Size:
8
9
10
Description
- Suitable for various applications and offer reliable protection against a wide range of chemicals and also mechanical hazards
- Ideal for: providing PPE Category 3 protection in many different segments like Aerospace, Automotive, Metal Manufacturing, Food, Transportation, Construction, when handling chemicals, oils, grease, alcohols, acids, bases or solvents also in immersion and providing the highest level of abrasion resistance
- Risk of linting is avoided protecting the products from contamination
- Longer length to protect wrist and lower forearm and allow full immersion
- With sand patch texturing on fingers and palm offering enhanced wet and dry grip
- AQL 0.65 for pinholes to provide superior protection, more productivity and efficiency
Specifications
Color | Green |
Material | Nitrile |
Product Type | Gloves |
Size | 9 |
Thickness (Metric) | 0.5 mm |
Dimensions (L x W) | 47.8 x 15.6cm |
Length (English) | 18.642 in. |
Length (Metric) | 47.8 cm |
Length (Metric) Cuff | 45 cm |
Weight (English) | 4.224 lb. |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'