missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Jenway™ Quartz Cuvettes

Brand:  Cole-Parmer™ 035056

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12712705

  • 334.00€ / Each


Estimated Shipment: 08-10-2021
Add to basket

Description

Description

Specifications

Specifications

7.0 mL
Quartz
Cuvette
20mm
20 mm
Videos
SDS
Documents
Certificates