Learn More
Verenigde Kunststof Bedrijven™ Polyethylene, Jerry Can Drain Valve
Click to view available options
Diameter (Metric) Inner Neck:
19 to 22 mm
Description
- HDPE Drain Valve
- Reusable
- Adjustable flow
- Easy opening and closing
- Mounted in Cap
- DIN 51
Specifications
Product Type | Spout |
Closure Material | Polyethylene |
Closure Type | Tap |
Diameter (Metric) Inner Neck | 19 to 22 mm |
Material | Polyethylene |
For Use With (Application) | Jerrican |