missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ JNJ 1013NC

Product Code. 30281233
Click to view available options
:
5 mg
This item is not returnable. View return policy

Product Code. 30281233

Brand: Tocris Bioscience™ 8030/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Negative control for the IRAK1 degrader JNJ 1013 (Cat. No. 8029)

JNJ 1013NC is the negative control for the IRAK1 degrader, JNJ 1013 (Cat. No. 8029).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(4-(4-(2-(((S)-1-((2S,4S)-4-Hydroxy-2-(((S)-1-(4-(4-methylthiazol-5-yl)phenyl)ethyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-2-oxoethoxy)piperidin-1-yl)-2-methoxyphenyl)-6-(1H-pyrazol-5-yl)picolinamide
CAS 2755075-91-1
Quantity 5 mg
Target Active Degraders
Molecular Formula C46H55N9O7S
Purity 0.95
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.