missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX JNK3/SAPK1b Protein, Active

Product Code. p-7228326
Encompass_Preferred
Click to view available options
Quantity:
10 μg
250 μg
Unit Size:
10µg
250µg
This item is not returnable. View return policy

Product Code. 17320254

Brand: Eurofins DiscoverX 14501

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Active, N-terminal His6 tagged, recombinant, full length human JNK3/SAPK1b, for use in Kinase Assays.

c-Jun N-terminal kinases (JNKs), a stress-activated protein kinase (SAPK), is involved in cellular responses to environmental stresses. JNK kinases can be activated by stimuli such as UV light, radiation, protein synthesis inhibitors, ceramide, DNA-damaging drugs, chemopreventive drugs, TNF-a, and interleukin 1. JNK binds to an NH2-terminal region of ATF2 and c-Jun and phosphorylates two sites within the activation domain of each transcription factor. Three JNK genes (Jnk1, -2 and -3) have been identified in humans; however, splice variants result in a total of 10 isoforms. Inhibition of JNK3 (MAPK10, Mitogen-activated protein kinase 10, Stress-activated protein kinase JNK3) may provide clinical benefit in diseases as diverse as immune inflammatory (joint, bowel, pulmonary), ischemic injury (brain, cardiac) and neurodegenerative (Alzheimer's, Parkinson's) diseases.

The JNK3 protein sequence is identical to the human JNK3/SAPK1b described inGenBank NM_138980. The DNA sequence, however, is more closely related to the rat cDNA described inGenBank NM_012806.

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl, pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Molecular Weight (g/mol) 53 kDa
Quantity 10 μg
Species Human
Recombinant Recombinant
Protein Tag N-terminal 6His
Expression System Recombinant enzyme expressed in Sf21 insect cells
Protein Form Full-length
Purity or Quality Grade ≥77%
Protein JNK3/SAPK1b, active
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.