missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ JW 67
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
JW 67 is an inhibitor of canonical Wnt pathway signaling (IC50 = 1.17 μM); targets the β-catenin destruction complex (GSK-3β/AXIN/APC) to induce β-catenin degradation. Selective for the canonical Wnt pathway over the Sonic hedgehog (Shh) and NF-κB pathways. Blocks G1/S cell cycle progression in colorectal cancer (CRC) cell lines (GI50 = 7.8 μM).
Specifications
Specifications
| Purity | >97% |
| Quantity | 50 mg |
| Product Type | JW 67 |
| Molecular Weight (g/mol) | 394.38 |
| Inhibitors | JW 67 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction