missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ JZL 195

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Dual inhibitor of fatty acid amide hydrolase (FAAH) and monoacylglycerol lipase (MAGL) (IC50 values are 2 and 4 nM respectively). Elevates anandamide and 2-arachidonoylglycerol levels in vivo. Shown to impair short-term memory in mice.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 433.46 |
| Purity | >98% |
| Molecular Weight (g/mol) | 433.464 |
| Chemical Name or Material | JZL 195 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction