man
Test
chevron-down
Select Language
English
French
Search
Order Status
Quick Order
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Specialty Lab Equipment, Instruments, and Apparatuses
Digestion Equipment
Digestors
13445539
Print
BUCHI K-415 Model TripleScrub Scrubber
Brand:
BUCHI 114152330
Additional Details :
Weight :
11.80000kg
Product Code.
13445539
7230.00€
/ Each
Quantity
Estimated Shipment: 21-06-2024
Log in
to see stock.
Specifications
Product Type
TripleScrub
Voltage
230 V
Dimensions (L x W x H)
526 x 347 x 417mm
Height (Metric)
417 mm
Width (Metric)
347 mm
Hertz
50/60 Hz
Depth (Metric)
526 mm
Electrical Requirements
230V, 50/60 Hz
Quantity
1 Ea.