Learn More
Anritsu™ Type K Magnet Probes
Product Code. p-8001095
Click to view available options
Temperature (Metric):
≤250°C
≤400°C
Style:
Miniature Round
Standard Square
Unit Size:
Each
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11932139
Brand: Anritsu™ MG22KGW1ANP
Only null left
Add to basket
Add to basket
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Designed with magnets to allow probes to be attached to measurement targets. Anritsu™ Type K Magnet Probes are temperature sensors designed for stationary surfaces.
  • Designed for stationary surface
  • Available with square or round footprint
  • Magnetic probe that attaches to steel surface
  • Thermocouple contact
Specifications
Accuracy ±1.4°C (100°C surface)
Analysis Time 2.0 sec.
Description Head size: 32L x 27W x 34mmD
Temperature (Metric) ≤400°C
Type Thermocouple K
Material Chromel-Alumel
Style Standard Square