missing translation for 'onlineSavingsMsg'
Learn More

Kanamycin Sulfate, Serva Electrophoresis™

Product Code. 11452214
Encompass_Preferred
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 11452214

Brand: Serva Electrophoresis 2689901

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Specifications

Physical Form Solid
pH 6.5 to 8.5
Melting Point 250°C
Odor Light
Color White
Quantity 1 g
Solubility Information Soluble in water
compliance-icons
Product Identifier
  • Kanamycin sulfate
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1
Hazard Statement
  • H360D-May damage the unborn child.
Precautionary Statement
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.