missing translation for 'onlineSavingsMsg'
Learn More

Nordvlies Katrin Classic Toilet 250

Product Code. 15740907
Click to view available options
Color:
White
Unit Size:
64 rolls
This item is not returnable. View return policy

Product Code. 15740907

Brand: Nordvlies 10477

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Classic Toilet 250 is quality conventional toilet paper.

  • Quality conventional toilet rolls
  • 2-Ply, white, 250 sheets
  • Soft, absorbent paper for maximum comfort
  • Suitable for all public environments
  • All Katrin toilet papers are made from high quality fibers and have been proven, via Dissolving test 90 seconds, Metsä Tissue 02 method, to break down and dissolve quickly and efficiently in water, under normal usage conditions
  • Certified under the Nordic Swan Eco Label
  • Certified under the EU Flower Eco Label
  • Dermatologically tested and approved

Specifications

Product Type Toilet Paper
Certifications/Compliance Nordic Swan Eco Label, EU Flower Eco Label
Color White
For Use With (Equipment) Katrin System Toilet Dispenser
Packaging Quantity 250 Sheets
Ply 2-ply
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.