Learn More
Cole-Parmer™ Keck™ Tubing Adapter
Shop All Cole-Parmer Products
Click to view available options
Diameter (Metric) Inner:
8 to 10 mm
Unit Size:
Each
Description
- Safely and easily connect tubing to glassware or other tubing, connect and disconnect with just a quarter twist
- Unique adapters eliminate broken glass ports and prevent twisted tubing
- Glassware adapters 06841-51 and -52 are sold separately
Specifications
Specifications
| Diameter (Metric) Inner | 8 to 10 mm |
| Packaging Quantity | 10 Adapters |
| Product Type | Tubing Adapter |