Learn More
DWK Life Sciences Kimble™ Kim-Kap™ Disposable Closures
Available in natural (clear) or choice of four colors
Brand:  DWK Life Sciences 73660-38
Product Code. 11770734
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
Available in natural (clear) or choice of four colors
  • For plain-end culture tubes
  • Keep tube contents sterile without need for cotton plugging
  • Polypropylene
  • Autoclavable
Caps for 38mm O.D. are designed for baffled culture flasks.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Closure
Polypropylene
38 mm O.D. Plain-end Culture Tubes
Autoclavable
Natural
Single-use