Learn More
DWK Life Sciences Kimble™ KIMAX™-51 Brand Reusable Serological Pipets with Tooled Opening
Product Code. 11878472
Click to view available options
Volume (Metric):
0.2 mL
1 mL
5 mL
10 mL
25 mL
Graduations:
0.01 mL
0.1 mL
Color:
Black
Blue
Orange
Red
White
Unit Size:
Pack of 12
Pack of 6
Product Code. 11878472
Brand: DWK Life Sciences 37034A25
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This item is not returnable. Afficher la politique du retour.
Tempered tip with permanent brown scale fused into uniform bore tubing without etchin
Informations sur la commande
Plugs not included.
Compliance
Calibrated To Deliver/Blow Out. Designed from ASTM™ E 1044.
TRUSTED_SUSTAINABILITY
Spécification
Certifications/Compliance ASTM E1044
Material Borosilicate Glass
Graduated Yes
Graduations 0.1 mL
Length (English) 15.74 in.
Tolerance ±0.1 mL
Color White
Volume (Metric) 25 mL
Length (Metric) 400 mm
Type Serological Pipet