Learn More
DWK Life Sciences Kimble™ Adapters for Round-Bottom Centrifuge Tubes
Product Code. 11843825
Click to view available options
Capacity:
15 mL Round-bottom Centrifuge Tube
30 mL Round-bottom Centrifuge Tube
Unit Size:
Pack of 2
Product Code. 11843825
Brand: DWK Life Sciences 4555030_OLD
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Required when centrifuging select Kimble high-strength round-bottom centrifuge tubes to 13,000xG in 50mL rotor
  • Made of rubber
TRUSTED_SUSTAINABILITY
Specifications
Material Rubber
Capacity 30 mL Round-bottom Centrifuge Tube