Learn More
DWK Life Sciences Kimble™ 2mL and 4mL Autosampler Vials without Closures: Crimp Openings
Click to view available options
Color:
Amber
Clear
Marking Spot:
No
Yes
Neck Style:
Regular
Wide
Unit Size:
Pack of 2000
Description
- O.D. x Length: 12 x 32mm
- Finish: Crimp
Order Info
100 vials/pack, 20 packs/case. Closures not included.
Specifications
Specifications
| Color | Amber |
| Diameter (Metric) Outer | 12 mm |
| Material | Borosilicate Glass |
| Volume (Metric) | 2.0 mL |
| For Use With (Application) | Chromatography applications |
| Height (English) | 1.25 in. |
| Neck Style | Wide |
| No. per Pack | 100 |
| Closure Type | Crimp Large Opening with Marking Spot |
| Height (Metric) | 32 mm |
| Show More |