Learn More
DWK Life Sciences Kimble™ KIMAX™ Flasks with standard taper Pennyhead Stoppers
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
2000 mL
Closure Size:
13
16
19
22
27
Height (English):
7.08 in.
9.84 in.
11.29 in.
13.3 in.
15.74 in.
Height (Metric):
18 cm
25 cm
28.7 cm
33.8 cm
40 cm
Unit Size:
Pack of 12
Pack of 4
Pack of 6
Description
Calibrated To Contain
- Sizes 5mL and larger have designated volumes within Class A tolerances
- Sizes 1 and 2mL are test-tube shaped
Compliance
Designed from requirements for Class A unserialized flasks of ASTM™ E 288
Specifications
Specifications
| Calibration | Calibrated To Contain |
| Closure Material | Glass |
| Capacity (Metric) | 2000 mL |
| Material | Borosilicate Glass |
| Height (English) | 15.74 in. |
| Height (Metric) | 40 cm |
| Tolerance | ±0.5 mL |
| Closure Size | 27 |