Learn More
DWK Life Sciences Kimble™ Kontes™ Gas Bubbler
Product Code. 11862923
Click to view available options
Diameter (Metric) Outside:
25 mm
Unit Size:
Each
Product Code. 11862923
Brand: DWK Life Sciences 6522200000
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Used with mineral oil to observe gaseous addition to reactions
  • May be used to vent a reaction mixture to the atmosphere
  • Tubing connections are of 8mm O.D.
TRUSTED_SUSTAINABILITY
Specifications
Type Gas Bubbler
Diameter (English) Outside 0.98 in.
Diameter (Metric) Outside 25 mm
Height (English) 7.28 in.
Height (Metric) 185 mm
Width (English) 3.14 in.
Width (Metric) 80 mm
For Use With (Equipment) Mineral oil to observe gaseous addition to reactions