Learn More
DWK Life Sciences Kimble™ Kontes™ Incubation Flask Stoppers and Center Well
Rubber stopper for flask sidearm provides a secure double seal
Brand:  DWK Life Sciences 882311-0000
Additional Details : Weight : 0.80000kg
Product Code. 11863173
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
Sidearm Flask Stoppers
  • Rubber stopper for flask sidearm provides a secure double seal
  • Sidearm stopper can be punctured with a syringe needle
  • Stoppers fit 10 and 25mL flasks
Order Info
Top and sidearm stoppers are not interchangeable.
Specifications
Specifications
Rubber
Off-center hole
144
Sleeved stopper