missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ KIMAX™ KimCote™ Heavy-Duty Serum Bottles
Click to view available options
Capacity (Metric):
4000 mL
9000 mL
Unit Size:
Each
Description
- KimCote™ plastic coating for added safety in the event of breakage
- Autoclavable
- Ultra-clear and extremely durable
- Resistant to many common laboratory chemicals
- Unique texture provides non-slip handling, wet or dry
Specifications
Specifications
| Capacity | 304.32 oz., 9000mL |
| Capacity (Metric) | 9000 mL |
| Capacity (English) | 304.32 oz. |
| Color | Clear |
| Diameter (English) | 7.99 in. |
| Material | Glass |
| Height (Metric) | 412 mm |
| Height (English) | 16.22 in. |
| Closure Material | Rubber |
| Closure Size | No. 8 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction