Learn More
DWK Life Sciences Kimble™ Kontes™ NMR Pipets
Extended-tip NMR pipets for retrieving and adding sample in NMR tubes
Brand: DWK Life Sciences 897085-0009
74.23 EUR valid until 2024-04-30
Use promo code "21637" to get your promotional price.
Additional Details : Weight : 0.80000kg
Description
Extended-tip NMR pipets for retrieving and adding sample in NMR tubes
- Available with 7 and 9 in. (17.7 and 23cm) tips for use with 7 and 9 in. long NMR tubes
Specifications
Borosilicate Glass | |
100/Cs. | |
NMR Pipet |