Learn More
DWK Life Sciences Kimble™ Closures for Sample Vials
Black phenolic closure with PTFE-lined cap
150.00€ - 404.00€
Specifications
Product Type | Screw Cap |
---|---|
Liner Material | PTFE, Rubber |
Material | Phenolic |
Color | Black |
Product Code | Brand | Closure Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Size | Price | Quantity & Availability | |||||
17781078
|
DWK Life Sciences
73802-18400 |
18-400 |
150.00€
Pack of 144 |
Estimated Shipment: 08-07-2024
Log in to see stock. |
|||||
12184001
|
DWK Life Sciences
73802-13425 |
13-425 |
169.00€
Pack of 144 |
Estimated Shipment: 08-07-2024
Log in to see stock. |
|||||
15365536
|
DWK Life Sciences
73802-15425 |
15-425 |
172.00€
Pack of 144 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
10701574
|
DWK Life Sciences
73802-20400 |
20-400 |
249.00€
Pack of 144 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
11853133
|
DWK Life Sciences
73802-24400 |
24-400 |
404.00€
Pack of 144 |
Estimated Shipment: 08-07-2024
Log in to see stock. |
|||||
Description
Black phenolic closure with PTFE-lined cap
- Works with media bottles and screw-thread vials
- Special black phenolic material (wood filled) very resistant to the effects of temperature and steam encountered in autoclaving
- Cemented-in PTFE-faced rubber liner is provided
Specifications
Screw Cap | |
PTFE, Rubber | |
Phenolic | |
Black |