Learn More
DWK Life Sciences Kimble™ Glass Racks for Westergren Tubes
Reduces the chance of bio hazardous contamination
595.00€
Product Code | Brand | Positions | For Use With (Application) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Positions | For Use With (Application) | Price | Quantity & Availability | ||||
15929665
|
DWK Life Sciences
2215N |
10 | For westergren and wintrobe tubes |
595.00€
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||||
Description
- Racks feature leveling bubbles and adjustment screws
- Tubes are held firmly in a perpendicular position to ensure accurate readings.
Specifications
Not Autoclavable | |
Glass |