Learn More
DWK Life Sciences Kimble™ KIMAX™ Conical-Bottom Glass Centrifuge Tubes, 10mL
Click to view available options
Disposable:
Reusable
Single-use
Material:
Borosilicate Glass
Heavy-duty Kimax Glass
Length (Metric):
108 mm
114 mm
116 mm
117 mm
Closure Type:
Not Included
Screw Cap
Unit Size:
Pack of 12
Pack of 125
Pack of 6
Description
Use these tubes for centrifugation up to 2980 RCF. KIMAX™ Conical-Bottom Glass Centrifuge Tubes are plastic shrink-wrapped in modular trays for cleanliness and safety. Manufactured from 51 expansion, low extractable borosilicate glass.
- With conical shaped bottom
- Tubes are made from borosilicate glass in compliance with ASTM Specification E438, Type I, Class A or B glass requirements
Specifications
Specifications
| Calibration | To contain |
| Certifications/Compliance | USP, ASTM E 438 |
| Capacity (Metric) | 10 mL |
| Disposable | Reusable |
| Material | Heavy-duty Kimax Glass |
| Liner Material | Rubber |
| Thread Size | 15-415 |
| Tolerance | 0 to 0.2mL in ±0.05mL, >0.2 to 1mL in ±0.1mL, >1 to 3mL in ±0.15mL, >3 in ±0.2mL |
| Shape | Conical |
| Length (Metric) | 116 mm |
| Show More |