Learn More
DWK Life Sciences Kimble™ Kontes™ Potter-Elvehjem Tissue Grinders
Product Code. 17730888
Click to view available options
Capacity:
0.13 oz., 4mL
0.33 oz., 10mL
1.01 oz., 30mL
Graduations:
0.5 to 4mL
1 to 9mL
2 to 25mL
Unit Size:
Each
Product Code. 17730888
Brand: DWK Life Sciences 8855100021
Only null left
Add to basket
Add to basket
With serrated PTFE pestle and graduated tube
  • Radial serrations on the lower portion of PTFE pestle deliver homogenate into cylindrical portion of tube
  • Stainless-steel pestle shaft
  • Graduations fused into the glass tube
  • Tubes and pestles are interchangeable
TRUSTED_SUSTAINABILITY
Specifications
Size 21
Diameter (English) Shaft 0.23 in.
Diameter (Metric) Shaft 6mm
Capacity (English) 0.33 oz.
Capacity (Metric) 10 mL
Product Type Pestle and Graduated Tube
Graduations 1 to 9mL
Length (English) Pestle 8.38 in.
Length (Metric) Pestle 213mm
Length (English) Tube 5.03 in.
Length (Metric) Tube 128mm
Capacity 0.33 oz., 10mL