missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Phenolic Caps with Pulp/Vinyl Liners

Product Code. p-3563954
Click to view available options
Neck Finish:
20-400
Unit Size:
Pack of 144
This item is not returnable. View return policy
Not recommended for hydrocarbons or bleaches.

Product Code. 15384052

Brand: DWK Life Sciences 75201G20400

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Not recommended for hydrocarbons or bleaches.

Cap provides good chemical resistance to mild acids, alkalis, alcohols, aqueous solutions, oils and solvents.

  • Polyvinyl film is adhered to 0.035-in. pulpboard
  • Color: black
  • Low cost cap for general purpose usage
  • Not recommended for hydrocarbons or bleaches

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Phenolic Resin
No. per Case 144 Cs.
Neck Finish 20-400
Product Type Phenolic Cap
For Use With (Equipment) Bottle
Closure Liner Pulp/Vinyl
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.