Learn More
Lonza™ Kinetic-QCL™ Bulk Kinetic Chromogenic LAL Assay; 2400 Test Kit
missing translation for 'orderingAttributeHoverText'
Quantity:
100 ViaLs LAL Lysate, 24 Tests/Vial
missing translation for 'unitSize'
Pack of 100
Description
- Bulk Kinetic Chromogenic LAL Assay for endotoxin detection for 2040 tests.
- Kit does not contain endotoxin: E50-643L is ordered separately.
- Available as a made-to-order product.
- Assay sensitivity: 0.005 - 50 EU/mL
- Most sensitive of the LAL assays.
- Quantitative, kinetic assay for the detection of Gram-negative bacterial endotoxin.
- A sample is mixed with the reconstituted LAL reagent in a 96-well plate and placed in an incubating plate reader that measures absorbance at 405 nm.
- The reaction is automatically monitored over time for the appearance of a yellow color.
- The time required for the change is inversely proportional to the amount of endotoxin present.
- The concentration of unknown samples can then be calculated from a standard curve.
- Most appropriate choice for testing small volume parenterals, vaccines, antibiotics and biologicals.
- Less impacted by inhibitory products that may interfere with the clotting mechanism in turbidimetric and gel clot assays.
- Ideal for biological products such as vaccines and antibiotics.
Specifications
Specifications
| Assays | Kinetic Chromogenic LAL Assay for endotoxin detection |
| Description | Assay sensitivity: 0.005 to 50 EU/mL |
| For Use With (Application) | Endotoxin detection |
| Quantity | 100 ViaLs LAL Lysate, 24 Tests/Vial |