Få mere at vide
Få mere at vide
Thermo Scientific™ KingFisher™ Cell and Tissue DNA Kit
Shop alle Thermo Scientific produkter
Klik for at se tilgængelige muligheder
Quantity:
96 Samples
Pakningsstørrelse:
Each
Beskrivelse
Rapidly purify high-quality, reproducible DNA from cell and tissue samples, as well as cultured bacteria, with the Thermo Scientific™ KingFisher™ Cell and Tissue DNA Kit. This unique nucleic acid purification workflow provides an optimized, high-throughput method for extreme flexibility.
Features:
- Purifies DNY from maximum 1 x 107 cells or 20mg of tissue or 1mL of cultured bacteria
- Produces isolated DNA that is free of inhibitors and ready for use in many downstream applications
- Includes easy-to-use protocols for safe sample-handling as well as minimal user input, maximizing walkaway time
- Optimized to produce high yields of quality DNA
- Suitable for wide range of sample materials from cells to tissues
- Flexible throughput from one sample to up to 96 samples per run
- Compatible with most common downstream analysis and applications
Recommended for:
Genomics, target identification, veterinary assays, biomarker discovery, quality control
Tekniske data
Tekniske data
| Sample Type | Tissue, Cell Culture, Bacteria |
| Suitable Downstream Analysis | PCR, Real-time PCR, Enzymatic Reactions |
| Description | KingFisher Cell and Tissue DNA Kit; 96 samples |
| Elution Volume (Metric) | 100µL (Duo) or 150µL (Flex) |
| Quantity | 96 Samples |
| Run Time | 25 min. |
| Sample Volume | Up to 20 mg of tissue, 1 x 107 cells, or 1 mL of overnight cultured bacteria |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion