Learn More
HUBER KISS 225B Heating Bath Circulator
Product Code. 15652717
Click to view available options
Voltage:
230 V
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 15652717
Brand: HUBER 2060.0004.98
Only null left
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
Heating bath circulators with a stainless steel bath allow the temperature control of internal and external thermoregulation tasks. Items can be directly thermoregulated by placing them in a thermostat bath and the control of externally connected applications is also possible.
  • Temperature of various objects can be controlled by placing them directly within bath
  • Fitted with overtemperature and low level protection suitable for Safety Class III/FL (DIN 12876) for use with flammable fluids
  • Pressure/suction pump ensures optimal circulation and temperature homogenity in bath
  • Allows temperature control of externally attached applications using pump adapter
  • KISS temperature controller
  • Simple operation with menu navigation
  • Simultaneous display of setpoint, Tmin, Tmax and actual temperature
  • Status displays for pump, cooling and heating
  • USB and RS232 interfaces
  • Autostart function for power failure
  • 3 colour versions available: grey (standard), blue, red
  • Made from stainless steel
  • Suitable for routine tasks in research and industry
Specifications
Controller Type KISS
Capacity (Metric) 25 L
Certifications/Compliance III/FL (DIN 12876)
Pump Type M16x1 AG Male
Temperature Range 25°C to 200°C
Type KISS 225 B Bath Thermostat with Stainless Steel Bath
Heating Capacity 2 kW
Material Stainless Steel
Max. Pressure 14 L/min; 0.25 bar
Model KISS 225 B
Temperature Stability ±0.05°C
Depth (Metric) 555 mm
Weight (English) 35.2 lb.
Weight (Metric) 16 kg
Voltage 230 V
Amperage 10 A
Frequency 50/60 Hz