Learn More
Invitrogen™ Human KIT [V559D T670I], GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Recombinant human KIT V559D T670I (544-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. , Validated in LanthaScreen™ Kinase Binding Assay, Protocols available for simple use and optimized performance, Multiple pack sizes available for convenient use Related Product: LanthaScreen™ Eu Kinase Binding Assay Protein Form: Recombinant
Specifications
Specifications
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5 |
| Molecular Weight (g/mol) | 75.3 kDa |
| Name | Human KIT [V559D T670I], GST Tag |
| Quantity | 10 μg |
| Common Name | KIT (V559D T670I Mutant) |
| Protein Length | 544-end |
| Conjugate | Unconjugated |
| Species | Human |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.