missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human KIT [V559D, V654A], GST Tag Recombinant Protein

Product Code. 15930283
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
100µg
10µg
1mg
This item is not returnable. View return policy

Product Code. 15930283

Brand: Invitrogen™ A30498

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This KIT V559D V654A recombinant human protein (544-end) was expressed in insect cells. KIT V559D V654A recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Z'-LYTE™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Recombinant, Active, Catalytic, Mutant The gene accession number for KIT [V559D is NP_000213.1.

KIT receptor tyrosine kinase is a proto-oncogene and a type 3 transmembrane receptor for MGF (mast cell growth factor, also known as stem cell factor). KIT was first identified as the cellular homolog of the feline sarcoma viral oncogene v-kit. KIT together with its ligand regulates growth and activation of a variety of hemopoietic and non-hemopoietic cells. Mutations in KIT are associated with gastrointestinal stromal tumors, mast cell disease, acute myelogenous leukemia, and piebaldism. Recently, deregulation of the KIT receptor TK by the prevalent activation loop mutation D816V has served as a focal point in therapeutic strategies aimed at curbing neoplastic mast cell growth.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P10721
Concentration See Label
For Use With (Application) Kinase Assay
Format Kit
Formulation 50mM tris HCl with 150mM NaCl, 0.1mM PMSF, 0.25mM DTT, 0.1mM EDTA, 10mM glutathione, 25% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 3815
Molecular Weight (g/mol) 75.2 kDa
Name Human KIT [V559D, V654A], GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias KIT
Common Name KIT (V559D, V654A Mutant)
Gene Symbol KIT
KinaseFamily PDGFR Kinase Family
Protein Length 544-end
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 544-end
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Form Recombinant, Active, Catalytic, Mutant
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.