missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ KLEENGUARD™ A10 Light Duty Coveralls with Hood

Offering CE Simple (minimal risk) protection for the user engaged in light duty tasks and basic contamination control. Kimberly-Clark™ KLEENGUARD™ A10 Light Duty Coveralls with Hood are manufactured from spunbond polypropylene material. 

Brand:  Kimberly-Clark™ 95660

Additional Details : Weight : 5.38560kg

 View more versions of this product

Product Code. 12671476

  • 258.00€ / Pack of 50


Estimated Shipment
01-03-2021
Add to basket

Description

Description

  • Silicone-free and individually packed
  • Reduces the risk of heat stress
  • Easy fastening snap front
  • Triple-stitched seams to help protect against tearing
  • Offers complete freedom of movement with comfortable body design and elasticated cuffs, ankles and waist
Specifications

Specifications

Elastic
Sterile
Blue
Yes
400 mm
400 mm
X-Large
Polypropylene
Elastic
Elastic
General maintenance, contract cleaning and construction utility areas
Yes
Stitched
300 mm
Videos
SDS
Documents