Learn More
Kimberly-Clark Professional™ KleenGuard™ Nemesis™ Safety Glasses
GREENER_CHOICE
Catalog Number p-7162434
Click to view available options
Frame Color:
Black
Lens Tint:
IR/UV 3.0, Shade 3
Lens Coating:
UV-treated
KleenGuard™ Nemesis™ Safety Glasses feature durable lightweight construction for protection at work and at home.
  • Sporty, flexible, lightweight design
  • Optional Anti-Fog coating prevents fogging; ideal for humid environments and temperature variances
  • Meets ANSI Z87.1+ standards for personal eye protection
  • Polycarbonate lenses provide 99.9% UVA/ UVB/UVC protection
  • Patented FLEX-DRY™ nosepiece is designed to be flexible, to channel sweat away from the eyes, and to help prevent slippage down the nose
  • Recyclable via The RightCycle Program by Kimberly-Clark Professional
  • Lens options include:
    • Smoke tint lenses with outer blue mirror finish coating is ideal for outdoor use where sunlight can cause eyestrain and fatigue
    • Copper blue shield lenses have a light brown tint and are designed to filter out blue light for enhanced sharpness and depth perception; ideal for low-light conditions
    • Bronze lenses provide visual definition with increased contrast and enhanced contours in most light conditions
    • Amber/yellow lenses block out blue light, making the environment appear brighter; ideal for low-light conditions and indoor use
    • IRUV Shade 3 lenses protect against infrared (IR) and ultraviolet radiation (UVA/UVB) during tasks like light cutting and brazing
    • IRUV Shade 5 lenses protect against infrared (IR) and ultraviolet radiation (UVA/UVB) during tasks like medium cutting brazing
    • Brown polarized lenses are effective for filtering scattered blue light to improve depth perception and contrast; ideal for outdoor use
TRUSTED_SUSTAINABILITY
Certifications/Compliance EN166:2001 1F for safety, EN166:2001 level F for impact resistance and EN166:2001 class 1 for optical clarity.
Lens Coating UV-treated
Lens Tint IR/UV 3.0, Shade 3
Product Type Safety Eyewear
For Use With (Application) Protecting eyes from sparks, spatter and small debris while working in welding or related areas, driving compliance through appealing styling
Frame Color Black
No. per Case 12 pairs/Cs.
No. per Pack 12/Pk.
Temple Style Soft touch
Weight (Metric) 0.324 kg
Lens Material Polycarbonate
Frame Material Plastic
UV Protection 0.999
Frame Type Wrap-around, IRUV 3.0 LENS