missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Incubation Flasks
For metabolic studies of biological materials
209.10€ - 335.58€
Specifications
Closure Material | Rubber |
---|---|
Capacity (Metric) | 10 mL |
Material | Borosilicate Glass |
For Use With (Application) | For measuring C14O2 yields following incubation, in-vitro incubations, Insulin bioassays, Persulfate oxidations and distillation of any volatile Acid or base |
Height (English) | 2 in. |
Product Code | Brand | Capacity (Metric) | Closure Material | Material | Includes | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Material | Material | Includes | Price | Quantity & Availability | ||
10208294
|
DWK Life Sciences
8823000010 |
10 mL | Rubber | Borosilicate Glass | w/o Sidearm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||
11843173
|
DWK Life Sciences
8823600010 |
10 mL | Rubber | Borosilicate Glass | Sidearm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||
Description
Ideal for measuring C14O2 yields following incubation, in vitro incubations, insulin bioassays, persulfate oxidations, and distillation of any volatile acid or base. Also suitable for use with phosphate or bicarbonate buffers when acid or alkali is introduced at the end of a reaction.
- Available with or without sidearm
- Flask with sidearm enables syringe needle entry without disturbing established conditions
Order Info
Stoppers and center well sold separately.
Specifications
Rubber | |
Borosilicate Glass | |
2 in. | |
1.2 in. | |
Stopper |
10 mL | |
For measuring C14O2 yields following incubation, in-vitro incubations, Insulin bioassays, Persulfate oxidations and distillation of any volatile Acid or base | |
5 cm | |
3.1 cm |