Learn More
DWK Life Sciences Kimble™ Kontes™ KIMAX™ Reusable High Strength Centrifuge Tubes
Product Code. 11875824
Click to view available options
Capacity (Metric):
15 mL
30 mL
Length (Metric):
102 mm
106 mm
Diameter (Metric):
18 mm
24 mm
Unit Size:
Pack of 6
Product Code. 11875824
Brand: DWK Life Sciences 4531050100
Only null left
Add to basket
Add to basket
Chemically strengthened to achieve a greater mechanical strength
  • Made of KIMAX Type 1, Class B, borosilicate glass
  • Can be centrifuged up to 13,100xG with rubber adapter sleeve
  • Withstand temperatures up to 300°C
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ASTM E438
Capacity (Metric) 30 mL
Disposable Reusable
Material Borosilicate Glass
Length (Metric) 106 mm
Capacity (English) 1.01 oz.
Diameter (Metric) 24 mm
Product Type Centrifuge Tube
Max. RCF 13,100 x g