Learn More
DWK Life Sciences Kimble™ Kontes™ Schlenk Vacuum Tube with HI-VAC Valve
Product Code. 11838532
Click to view available options
Overall Length (English):
4.72 in.
5.9 in.
6.49 in.
Width (English):
2.55 in.
2.75 in.
2.95 in.
3.14 in.
Height (English):
3.93 in.
4.72 in.
5.9 in.
Unit Size:
Each
Product Code. 11838532
Brand: DWK Life Sciences 2187100050
Only null left
Add to basket
Add to basket
Suitable for use with K216050 or K216060 Manifolds
  • Medium wall borosilicate glass
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
Material Borosilicate Glass
Diameter (English) Outer 1.25 in.
Diameter (Metric) Outer 3.2 cm
Overall Length (English) 4.72 in.
Overall Length (Metric) 12 cm
For Use With 216050 or 216060 manifolds
Width (Metric) 7.5 cm
Width (English) 2.95 in.
Height (Metric) 12 cm
Height (English) 4.72 in.
Includes Hi-Vac™ valve, valve plug
Product Type Vacuum Tube