Learn More
IKA™ KS 4000 i Thermostable Shaker
GREENER_CHOICE
Product Code. 10408643 Shop All IKA Products
Click to view available options
Speed Range:
10 to 500 rpm
Unit Size:
Each
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 10408643
Brand: IKA™ 0003510100
Only null left
Add to basket
Add to basket
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Specifications
Speed Range 10 to 500 rpm
Electrical Requirements 230/115/100 V, 50/60 Hz, 1120 W
Voltage 100/115/230 V
Temperature Range (Metric) -10°C to +80°C
Wattage 1120 W
Holds 90L
Product Type Thermostable Shaker
Certifications/Compliance IP 30 DIN EN 60529
Frequency 50/60 Hz
Depth (Metric) 750 mm
Height (Metric) 525 mm
Width (Metric) 580 mm
Weight (Metric) 55 kg
Model KS 4000 ic control
Display LED
Humidity Range 80%
Interface RS-232
Warranty and Service Offering 24 Months