Learn More
Heidolph™ Kugelmeiers 3D Cell Culture Plate SP5D (Sphericalplate 5D)
Shop All Heidolph Instruments Products
Click to view available options
Array:
2 x 3
4 x 6
No. of Wells:
6
24
Unit Size:
Each
Beskrivning
The patented geometry with special coating allows the integration of every single cell into a 3D-agglomerate in a controlled way. This is how uniformly sized spheroids grow without the formation of necrotic cores. The reproducibility of follow-up tests increases, since the experiments always start under identical initial conditions due to the low size variance of the cultivated spheroids.
- According to the size and scale of the experiments, you can choose between our 24-well plate or our 6-well plate
- For small batches, Heidolph™ suggests using the 24-well plate containing 12 wells with 750 microwells each so you will be able to yield up to 9000 uniform spheroids
- You can comfortably carry out monolayer comparative studies in the remaining 12 wells of the same plate
- 6-well plate is suitable for scale-up trials and bigger batches; you can expect a yield of 3364 spheroids per well, thus a total yield of 20,184 spheroids/plate
- The ready to use SP5D is particularly user-friendly: The cultivation does not require any pretreatment or centrifugation steps after seeding your cells
- The media change is also particularly convenient due to simple pipetting, since the height of the microwells was chosen so that the cell clusters are retained
- Platform is compatible with standard automation units
Specifikationer
Specifikationer
| Barcode | No Barcode |
| Format | Standard |
| Lid | With Lid |
| Array | 4 x 6 |
| Autoclavable | Not Autoclavable |
| Color | Transparent |
| Disposable | Single-use |
| For Use With (Equipment) | Standard Automation Unit |
| Material | Cycloolefin Copolymer |
| No. of Wells | 24 |
| Visa mer |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering