missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Alanine Benzyl Ester Hydrochloride, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | L-Alanine Benzyl Ester Hydrochloride |
| CAS | 5557-83-5 |
| Molecular Formula | C10H14ClNO2 |
| Quantity | 25 g |
| Synonym | L-Alanine Phenylmethyl Ester Hydrochloride, (S)-Benzyl 2-aminopropanoate Hydrochloride, Benzyl L-Alaninate Hydrochloride, NSC 523831 |
| InChI Formula | InChI=1S/C10H13NO2.ClH/c1-8(11)10(12)13-7-9-5-3-2-4-6-9;/h2-6,8H,7,11H2,1H3;1H/t8-;/m0./s1 |
| SMILES | Cl.C[C@H](N)C(=O)OCc1ccccc1 |
| IUPAC Name | benzyl (2S)-2-aminopropanoate;hydrochloride |
| Molecular Weight (g/mol) | 215.68 |
| Formula Weight | 215.0713 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction