Learn More
2-n-Propylphenol, 97%, Thermo Scientific™
Product Code. 11458953
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11458953
Brand: Thermo Scientific Alfa Aesar A11088.06
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 644-35-9
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00002251
InChI Key LCHYEKKJCUJAKN-UHFFFAOYSA-N
Synonym 2-n-propylphenol, o-propylphenol, phenol, 2-propyl, phenol, o-propyl, 1-hydroxy-2-n-propylbenzene, 1-2-hydroxyphenyl propane, propylphenol, 2-propyl-phenol, o-n-propylphenol, unii-333r6f6t3n
PubChem CID 12570
IUPAC Name 2-propylphenol
SMILES CCCC1=CC=CC=C1O
Specifications
Density 0.989
Boiling Point 214°C
Flash Point 93°C (199°F)
Odor Phenol-like
Refractive Index 1.528
Quantity 5 g
UN Number UN3145
Beilstein 1363932
Formula Weight 136.19
Percent Purity 97%
Chemical Name or Material 2-n-Propylphenol