Learn More
L-Asp-Asp-rhodamine Trifluoroacetate Salt, Thermo Scientific™
Click to view available options
Quantity:
0.5 mg
1 mg
Unit Size:
0.5mg
1mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Color | White to Off-white |
| Product Type | Detergent |
| Molecular Formula | C28H25N4O9·CF3CO2H |
| Formula Weight | 788.56g/mol |
| Synonym | (Asp)2-R110; Rhodamine 110, bis-(L-aspartic acid amide), trifluoroacetic acid salt |
| Sensitivity | Light Sensitive |
| Content And Storage | -30°C to -10°C |
| Form | Lyophilized |
| Quantity | 0.5 mg |