Learn More
L-Aspartic acid monosodium salt monohydrate, 97%, Acros Organics
Marque: Acros Organics 451620010
Informations supplémentaires : CAS : 323194-76-9 Poids : 1.00000kg
Quantité. | 1kg |
---|---|
Emballage | Plastic bottle |
Identifiants chimiques
3792-50-5 | |
192.123 | |
monosodium l-aspartate dihydrate | |
(2S)-2-aminobutanedioic acid;sodium;dihydrate |
C4H11NNaO6 | |
VJDUMAWJHIHDHJ-SQGDDOFFSA-N | |
129893661 | |
C(C(C(=O)O)N)C(=O)O.O.O.[Na] |
Spécification
L-Aspartic acid monosodium salt monohydrate | |
97% | |
monosodium l-aspartate dihydrate | |
VJDUMAWJHIHDHJ-SQGDDOFFSA-N | |
(2S)-2-aminobutanedioic acid;sodium;dihydrate | |
129893661 | |
97% |
3792-50-5 | |
C4H11NNaO6 | |
Plastic bottle | |
C(C(C(=O)O)N)C(=O)O.O.O.[Na] | |
192.123 | |
173.1 | |
1kg |