Learn More
L-Bornyl acetate, 95%, Thermo Scientific Chemicals
Catalog Number p-72149
20.86 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
5655-61-8
C12H20O2
196.29
MFCD00867808,MFCD00135942,MFCD00135942
KGEKLUUHTZCSIP-HOSYDEDBSA-N
bornyl acetate, -bornyl acetate
44630108
[(1R,3S)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] acetate
CC(=O)O[C@@H]1C[C@@H]2CC[C@@]1(C)C2(C)C
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
5655-61-8
C12H20O2
196.29
MFCD00867808,MFCD00135942,MFCD00135942
KGEKLUUHTZCSIP-HOSYDEDBSA-N
bornyl acetate, -bornyl acetate
44630108
[(1R,3S)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] acetate
CC(=O)O[C@@H]1C[C@@H]2CC[C@@]1(C)C2(C)C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.9875g/mL
Boiling Point 223°C to 224°C
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (as C12H20O2) Total esters (GC)
Packaging Glass bottle
Refractive Index 1.4620 to 1.465
Quantity 5 g
Beilstein 06,82
Merck Index 15,1341
Specific Gravity 0.9875
Specific Rotation -38
Solubility Information Solubility in water: .. Other solubilities: miscible with 95% alcohol and ether
Formula Weight 196.29
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material L-Bornyl acetate, 95%