Learn More
L-(-)-Glucose, anhydrous, 98%, Thermo Scientific Chemicals
Description
L-(-)-Glucose is an enantiomer of the more common D-glucose a naturally occurring carbohydrate used in an abundance of cellular processes. L-Glucose is a synthetic sugar used in the formation of L-Glucose Pen taacetate, a potential therapeutic agent regarding type II diabetes. In addition, L-glucose can be used as a colon cleansing agent for before a colonoscopy procedure.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
L-(-)-Glucose is an enantiomer of the more common D-glucose a naturally occurring carbohydrate used in an abundance of cellular processes. L-Glucose is a synthetic sugar used in the formation of L-Glucose Pen taacetate, a potential therapeutic agent regarding type II diabetes. In addition, L-glucose can be used as a colon cleansing agent for before a colonoscopy procedure.
Solubility
Soluble in water, alcohol.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Melting Point | 148°C to 157°C |
| Quantity | 1 g |
| Beilstein | 1724626 |
| Synonym | l-glucose, l---glucose, levoglucose, l--glucose, 2s,3r,4s,5s-2,3,4,5,6-pentahydroxyhexanal, aldehydo-l-glucose, aldehydo-l-gluco-hexose, d-glucose-13c6,d7, levoglucose usan, glucose, l-isomer |
| Solubility Information | Soluble in water,alcohol. |
| InChI Key | GZCGUPFRVQAUEE-VANKVMQKSA-N |
| Optical Rotation | −53° (c=10 in Water, 24h) |
| SMILES | C(C(C(C(C(C=O)O)O)O)O)O |
| IUPAC Name | (2S,3R,4S,5S)-2,3,4,5,6-pentahydroxyhexanal |
| Molecular Weight (g/mol) | 180.156 |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.