missing translation for 'onlineSavingsMsg'
Learn More

R&DSystems™ L-Glutamine, powder

Product Code. 18482250
Click to view available options
Quantity:
100 gm
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 18482250

Brand: R&DSystems™ R90010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Formula: C5H10N2O3

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room temperature, under dry conditions
For Use With (Application) Cell Culture
Quantity 100 gm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.