Learn More
L-Phenylalanine methyl ester hydrochloride, 98%, Acros Organics
Brand:  Acros Organics 130322500
Additional Details : CAS : 7524-50-7 Weight : 0.25000kg
Packaging Glass bottle
Quantity 250g
 View more versions of this product
Product Code. 15109183
  • 602.30€ / 250g
Estimated Shipment date
from Supplier 17-06-2020
Add to basket
Specifications
Chemical Identifiers
7524-50-7
215.68
SWVMLNPDTIFDDY-FVGYRXGTSA-N
75736
COC(=O)C(CC1=CC=CC=C1)N.Cl
C10H13NO2·HCl
MFCD00012489
l-phenylalanine methyl ester hydrochloride, h-phe-ome.hcl, methyl l-phenylalaninate hydrochloride, s-methyl 2-amino-3-phenylpropanoate hydrochloride, unii-47hk4y94ja, h-phe-ome hcl, h-phe-ome hydrochloride, l-phenylalanine, methyl ester, hydrochloride, l-phenylalanine methyl ester hcl
methyl (2S)-2-amino-3-phenylpropanoate;hydrochloride
Specifications
L-Phenylalanine methyl ester hydrochloride
Glass bottle
White
250g
7524-50-7
C10H13NO2·HCl
MFCD00012489
l-phenylalanine methyl ester hydrochloride, h-phe-ome.hcl, methyl l-phenylalaninate hydrochloride, s-methyl 2-amino-3-phenylpropanoate hydrochloride, unii-47hk4y94ja, h-phe-ome hcl, h-phe-ome hydrochloride, l-phenylalanine, methyl ester, hydrochloride, l-phenylalanine methyl ester hcl
COC(=O)C(CC1=CC=CC=C1)N.Cl
215.68
215.68
98%
Authentic
+ 37.00
156.0°C to 160.0°C
98%
97.5% min. (Argentometry)
C6H5CH2CH(NH2)CO2CH3·HCl
14,499
SWVMLNPDTIFDDY-FVGYRXGTSA-N
methyl (2S)-2-amino-3-phenylpropanoate;hydrochloride
75736
Fine Crystalline Powder