Learn More
Berkshire™ Lab-Tips™ Low TOC Swab Large
Product Code. 17151359 Shop All Berkshire Products
Click to view available options
Length (Metric):
100 mm
Unit Size:
Pack of 25
Product Code. 17151359
Brand: Berkshire™ LTP125TOC.25
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Berkshire Lab-Tips™ Low TOC Swab Large knitted polyester swab features a unique semi-flexible internal paddle for greater surface coverage.
  • Specifically designed for Total Organic Carbon analysis and TOC tests in low-TOC environment
  • Critically low particles, fibers, ions and extractables
  • Excellent chemical resistance
  • High abrasion resistance while soft and non-abrasive
  • No contaminating adhesives and good sorbency
Specifications
Quantity 20 swabs/pack; 25 packs/case
Length (English) 3.94 in.
Product Type TOC Swab
Length (Metric) 100 mm
Material Polyester
Size Large